Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTCTACCTGATGCTGGACCGGGG[A/T]CACTTAGACTACCCCAGGAACCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 154582 | ||||||||||||||||||||
Literature Links: |
MAN2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAN2A1 - mannosidase alpha class 2A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002372.3 | 578 | Silent Mutation | GGA,GGT | G,G 29 | NP_002363.2 | |
XM_011543395.2 | 578 | Silent Mutation | GGA,GGT | G,G 29 | XP_011541697.1 | |
XM_017009472.1 | 578 | Intron | XP_016864961.1 |