Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGTGATCTCGCACCAGCCCTG[C/T]TCCCACAGGCCTGGCAGGAGGTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608005 | ||||||||||||||||||||
Literature Links: |
SIL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIL1 - SIL1 nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037633.1 | 1368 | Silent Mutation | GAA,GAG | E,E 375 | NP_001032722.1 | |
NM_022464.4 | 1368 | Silent Mutation | GAA,GAG | E,E 375 | NP_071909.1 | |
XM_011543570.2 | 1368 | Silent Mutation | GAA,GAG | E,E 385 | XP_011541872.1 |