Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAGTATGAACAAAGTTATAGGA[C/T]ACTACAAACAGTTGGTAATATATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM173B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM173B - family with sequence similarity 173 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258388.1 | 1120 | UTR 3 | NP_001245317.1 | |||
NM_001258389.1 | 1120 | UTR 3 | NP_001245318.1 | |||
NM_199133.3 | 1120 | UTR 3 | NP_954584.2 | |||
XM_011513964.1 | 1120 | UTR 3 | XP_011512266.1 |