Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCGAGTCTTCCCCTCCCCTCAG[C/G]CAGAAGCCGACGCCAACCCCGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LVRN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LVRN - laeverin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173800.4 | 367 | Silent Mutation | AGC,AGG | S,R 63 | NP_776161.3 | |
XM_011543263.2 | 367 | Silent Mutation | AGC,AGG | S,R 63 | XP_011541565.1 | |
XM_011543264.2 | 367 | Silent Mutation | AGC,AGG | S,R 63 | XP_011541566.1 | |
XM_017009215.1 | 367 | Silent Mutation | AGC,AGG | S,R 63 | XP_016864704.1 |