Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGAACCGGGTGGTGCTGCTC[C/T]ACGGTAAATTATGCAGGCTTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD9 - bromodomain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIP13 - thyroid hormone receptor interactor 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166260.1 | 677 | Missense Mutation | CAC,TAC | H,Y 178 | NP_001159732.1 | |
NM_004237.3 | 677 | Missense Mutation | CAC,TAC | H,Y 178 | NP_004228.1 | |
XM_011514163.2 | 677 | Missense Mutation | CAC,TAC | H,Y 178 | XP_011512465.1 |