Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATATTTTCTTTTTTTTTTACCTT[C/G]TCCAAATTTTGTTTTCTCAAACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD31 - ankyrin repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164443.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1825 | NP_001157915.1 | |
XM_011543298.2 | 5717 | Missense Mutation | CAA,GAA | Q,E 1883 | XP_011541600.1 | |
XM_011543299.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1882 | XP_011541601.1 | |
XM_011543300.2 | 5717 | Intron | XP_011541602.1 | |||
XM_011543301.2 | 5717 | Missense Mutation | CAA,GAA | Q,E 1826 | XP_011541603.1 | |
XM_011543302.2 | 5717 | Missense Mutation | CAA,GAA | Q,E 1767 | XP_011541604.1 | |
XM_017009317.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1850 | XP_016864806.1 | |
XM_017009318.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1836 | XP_016864807.1 | |
XM_017009319.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1786 | XP_016864808.1 | |
XM_017009320.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1767 | XP_016864809.1 | |
XM_017009321.1 | 5717 | Missense Mutation | CAA,GAA | Q,E 1449 | XP_016864810.1 | |
XM_017009322.1 | 5717 | Intron | XP_016864811.1 |