Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGTATGTTCCCTTTTGCCTTACT[A/G]TATGTTCTGTCAGTTTCTTTCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607003 | ||||||||||||||||||||
Literature Links: |
TSLP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSLP - thymic stromal lymphopoietin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033035.4 | 220 | Silent Mutation | CTA,CTG | L,L 7 | NP_149024.1 | |
NM_138551.4 | 220 | Intron | NP_612561.2 | |||
XM_011543698.1 | 220 | Intron | XP_011542000.1 |