Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTACTGTTTTTGTGAATTAACAG[A/C]GACCACCTTTCATGCCTCCTCCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCERG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCERG1 - transcription elongation regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040006.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | NP_001035095.1 | |
NM_006706.3 | 512 | Silent Mutation | AGA,CGA | R,R 96 | NP_006697.2 | |
XM_005268365.2 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_005268422.1 | |
XM_017008977.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_016864466.1 | |
XM_017008978.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_016864467.1 | |
XM_017008979.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_016864468.1 | |
XM_017008980.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_016864469.1 | |
XM_017008981.1 | 512 | Intron | XP_016864470.1 | |||
XM_017008982.1 | 512 | UTR 5 | XP_016864471.1 | |||
XM_017008983.1 | 512 | UTR 5 | XP_016864472.1 | |||
XM_017008984.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_016864473.1 | |
XM_017008985.1 | 512 | Silent Mutation | AGA,CGA | R,R 96 | XP_016864474.1 |