Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCAAATGACTCCCCAGTACCT[A/G]ACAGATGGATGTGAACATGAAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS14 - ribosomal protein S14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025070.1 | 1626 | Intron | NP_001020241.1 | |||
NM_001025071.1 | 1626 | Intron | NP_001020242.1 | |||
NM_005617.3 | 1626 | Intron | NP_005608.1 | |||
XM_017009684.1 | 1626 | UTR 3 | XP_016865173.1 |