Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAGCATTGTAGTGGCCACCCT[C/T]GTTCATCAGAATCTTGTCAAACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1QTNF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1QTNF2 - C1q and tumor necrosis factor related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031908.4 | 688 | Missense Mutation | AAG,GAG | K,E 221 | NP_114114.2 | |
XM_011534425.2 | 688 | Missense Mutation | AAG,GAG | K,E 260 | XP_011532727.1 | |
XM_011534426.2 | 688 | Missense Mutation | AAG,GAG | K,E 192 | XP_011532728.1 | |
XM_011534427.2 | 688 | UTR 3 | XP_011532729.1 | |||
XM_017009013.1 | 688 | Missense Mutation | AAG,GAG | K,E 153 | XP_016864502.1 |
CCNJL - cyclin J like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |