Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCCTTTTGGTGCTGGAGAGTC[C/T]CTGAGGGAGAGAAGTTTGACGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601798 MIM: 605675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNPDA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNPDA1 - glucosamine-6-phosphate deaminase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005471.4 | 1623 | UTR 3 | NP_005462.1 | |||
XM_005268348.1 | 1623 | UTR 3 | XP_005268405.1 | |||
XM_006714747.2 | 1623 | UTR 3 | XP_006714810.1 | |||
XM_011537537.1 | 1623 | UTR 3 | XP_011535839.1 |
RNF14 - ring finger protein 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |