Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCATTACAGCCACCATTCGGCC[C/T]AGACTTCTACTCAAGTGACACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610572 MIM: 603139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARVELD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARVELD2 - MARVEL domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038603.2 | 338 | Missense Mutation | CCA,CTA | P,L 59 | NP_001033692.2 | |
NM_001244734.1 | 338 | Missense Mutation | CCA,CTA | P,L 59 | NP_001231663.1 | |
XM_005248445.4 | 338 | Missense Mutation | CCA,CTA | P,L 59 | XP_005248502.1 | |
XM_005248446.4 | 338 | Missense Mutation | CCA,CTA | P,L 59 | XP_005248503.1 | |
XM_005248447.4 | 338 | Missense Mutation | CCA,CTA | P,L 59 | XP_005248504.1 |
RAD17 - RAD17 checkpoint clamp loader component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |