Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACAGCCTGTCAGATGCTGCCGG[C/G]GTCGGAGTCGACGGGCCCAACGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01184 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01184 - long intergenic non-protein coding RNA 1184 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A2 - solute carrier family 12 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001046.2 | 647 | Silent Mutation | GGC,GGG | G,G 161 | NP_001037.1 | |
NM_001256461.1 | 647 | Silent Mutation | GGC,GGG | G,G 161 | NP_001243390.1 | |
XM_011543588.2 | 647 | Silent Mutation | GGC,GGG | G,G 161 | XP_011541890.1 | |
XM_017009771.1 | 647 | Intron | XP_016865260.1 |