Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAACAGGTCTGAAATCTTTGGA[A/T]CTCTCGCGCAACTCCTTGGTTAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616475 | ||||||||||||||||||||
Literature Links: |
CEP72 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP72 - centrosomal protein 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018140.3 | 198 | Missense Mutation | GAA,GAT | E,D 60 | NP_060610.2 | |
XM_005248322.3 | 198 | Missense Mutation | GAA,GAT | E,D 10 | XP_005248379.1 | |
XM_011514063.1 | 198 | Missense Mutation | GAA,GAT | E,D 60 | XP_011512365.1 | |
XM_011514064.2 | 198 | Missense Mutation | GAA,GAT | E,D 10 | XP_011512366.1 | |
XM_017009626.1 | 198 | UTR 5 | XP_016865115.1 | |||
XM_017009627.1 | 198 | UTR 5 | XP_016865116.1 |
LOC100996325 - uncharacterized LOC100996325 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |