Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGTCTAGATTTTCGAGCTAATC[A/G]TCCCTCCAGCCCTCTGTGATCTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603640 MIM: 609383 | ||||||||||||||||||||
Literature Links: |
ADAM19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM19 - ADAM metallopeptidase domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033274.4 | 2082 | Missense Mutation | ACG,ATG | T,M 913 | NP_150377.1 | |
XM_005266003.2 | 2082 | Intron | XP_005266060.1 | |||
XM_011534682.2 | 2082 | Missense Mutation | ACG,ATG | T,M 646 | XP_011532984.1 | |
XM_017010009.1 | 2082 | Missense Mutation | ACG,ATG | T,M 646 | XP_016865498.1 |
NIPAL4 - NIPA like domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |