Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCTGCTGGCAGCAGTGGCTCCGG[A/C]GCCACCTCATCCAGCAGGGGCACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126660 | ||||||||||||||||||||
Literature Links: |
DBN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBN1 - drebrin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004395.3 | 1836 | Silent Mutation | GCG,GCT | A,A 516 | NP_004386.2 | |
NM_080881.2 | 1836 | Silent Mutation | GCG,GCT | A,A 518 | NP_543157.1 | |
XM_005265827.3 | 1836 | Silent Mutation | GCG,GCT | A,A 562 | XP_005265884.1 | |
XM_006714826.2 | 1836 | Silent Mutation | GCG,GCT | A,A 499 | XP_006714889.1 | |
XM_011534446.2 | 1836 | Silent Mutation | GCG,GCT | A,A 564 | XP_011532748.1 | |
XM_011534447.2 | 1836 | Silent Mutation | GCG,GCT | A,A 551 | XP_011532749.1 | |
XM_017009139.1 | 1836 | Silent Mutation | GCG,GCT | A,A 564 | XP_016864628.1 | |
XM_017009140.1 | 1836 | Silent Mutation | GCG,GCT | A,A 518 | XP_016864629.1 |
PRR7 - proline rich 7 (synaptic) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |