Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAGCTGTTAAGTTGGGGAAAAA[C/G]ATTTTTCCATACTTTTCCACATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NME5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NME5 - NME/NM23 family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003551.2 | 200 | Missense Mutation | ATC,ATG | I,M 65 | NP_003542.1 | |
XM_005272099.1 | 200 | Missense Mutation | ATC,ATG | I,M 65 | XP_005272156.1 | |
XM_006714716.3 | 200 | Missense Mutation | ATC,ATG | I,M 65 | XP_006714779.1 | |
XM_017009945.1 | 200 | Missense Mutation | ATC,ATG | I,M 65 | XP_016865434.1 |