Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGCCTACTACCCAGTGACTGT[C/G]AAAAAACAATTACGGGCCCAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCCC2 - methylcrotonoyl-CoA carboxylase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022132.4 | 588 | Silent Mutation | GTC,GTG | V,V 150 | NP_071415.1 | |
XM_005248567.1 | 588 | Silent Mutation | GTC,GTG | V,V 150 | XP_005248624.1 | |
XM_011543529.2 | 588 | Silent Mutation | GTC,GTG | V,V 150 | XP_011541831.1 | |
XM_017009688.1 | 588 | Silent Mutation | GTC,GTG | V,V 150 | XP_016865177.1 |