Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCACGGTCTCAAAGTTGTCCCCG[C/T]GAGCCAGCGCCTCGGCCTCGGCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MBLAC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBLAC2 - metallo-beta-lactamase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203406.1 | 36 | Missense Mutation | CAC,CGC | H,R 110 | NP_981951.1 |
POLR3G - RNA polymerase III subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006467.2 | 36 | Intron | NP_006458.2 | |||
XM_005248404.3 | 36 | Intron | XP_005248461.1 | |||
XM_011543100.2 | 36 | UTR 5 | XP_011541402.1 | |||
XM_011543101.2 | 36 | Intron | XP_011541403.1 | |||
XM_011543102.2 | 36 | Intron | XP_011541404.1 | |||
XM_011543103.2 | 36 | Intron | XP_011541405.1 | |||
XM_011543104.2 | 36 | Intron | XP_011541406.1 | |||
XM_011543105.2 | 36 | Intron | XP_011541407.1 | |||
XM_017008957.1 | 36 | Intron | XP_016864446.1 |