Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATTTAATAATCTTACAGCCTT[A/G]CATCCAAGGGTCCTTAAGCCGTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610889 | ||||||||||||||||||||
Literature Links: |
IPO11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IPO11 - importin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134779.1 | 1259 | Intron | NP_001128251.1 | |||
NM_016338.4 | 1259 | Intron | NP_057422.3 |
IPO11-LRRC70 - IPO11-LRRC70 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC70 - leucine rich repeat containing 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181506.4 | 1259 | Silent Mutation | TTA,TTG | L,L 340 | NP_852607.3 |