Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACATGTTTCAAAACCAAGAGTT[A/C]TAAGAAATGAAAAAGAGATATGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTX3 - metaxin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THBS4 - thrombospondin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306212.1 | Intron | NP_001293141.1 | ||||
NM_001306213.1 | Intron | NP_001293142.1 | ||||
NM_001306214.1 | Intron | NP_001293143.1 | ||||
NM_003248.5 | Intron | NP_003239.2 | ||||
XM_017009798.1 | Intron | XP_016865287.1 | ||||
XM_017009799.1 | Intron | XP_016865288.1 | ||||
XM_017009800.1 | Intron | XP_016865289.1 |