Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCTGGATGAAGAGTTTTCGCA[A/T]CTCCTCTGGGGACTTAGATGGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138491 | ||||||||||||||||||||
Literature Links: |
GLRA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRA1 - glycine receptor alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000171.3 | 1497 | Missense Mutation | ATG,TTG | M,L 404 | NP_000162.2 | |
NM_001146040.1 | 1497 | Missense Mutation | ATG,TTG | M,L 412 | NP_001139512.1 | |
NM_001292000.1 | 1497 | Missense Mutation | ATG,TTG | M,L 321 | NP_001278929.1 | |
XM_017009349.1 | 1497 | Missense Mutation | ATG,TTG | M,L 420 | XP_016864838.1 |