Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTTACGGTCCCTCTGGCCAGT[A/T]CACCCATGAATTTGATGGAGATGAG
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 146880 | |||||||||||||||||||||||||||||
Literature Links: |
HLA-DQA1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
AFR
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
SAS - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
HLA-DQA1 - major histocompatibility complex, class II, DQ alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002122.3 | 199 | Missense Mutation | TAC,TTC | Y,F 48 | NP_002113.2 | |
XM_006715079.3 | 199 | Missense Mutation | TAC,TTC | Y,F 48 | XP_006715142.1 |