Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCTCTGCCGGAAGCCCTGTTC[C/T]GAGGAGGTGCTAGGGACAGGCTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM40 - tripartite motif containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286633.1 | 1000 | Silent Mutation | TCC,TCT | S,S 61 | NP_001273562.1 | |
NM_138700.4 | 1000 | Silent Mutation | TCC,TCT | S,S 61 | NP_619645.1 | |
XM_011514305.1 | 1000 | Silent Mutation | TCC,TCT | S,S 61 | XP_011512607.1 | |
XM_011514306.1 | 1000 | Silent Mutation | TCC,TCT | S,S 61 | XP_011512608.1 | |
XM_011514308.1 | 1000 | Silent Mutation | TCC,TCT | S,S 61 | XP_011512610.1 | |
XM_011514309.1 | 1000 | Silent Mutation | TCC,TCT | S,S 61 | XP_011512611.1 |