Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGTAGCTGGGCCCGGAGGTAGC[A/G]GAGGTCTTGCTGATCAAGCCCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRM - nurim (nuclear envelope membrane protein) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270707.1 | 1070 | Missense Mutation | CGC,TGC | R,C 246 | NP_001257636.1 | |
NM_001270708.1 | 1070 | UTR 3 | NP_001257637.1 | |||
NM_001270709.1 | 1070 | Missense Mutation | CGC,TGC | R,C 181 | NP_001257638.1 | |
NM_001270710.1 | 1070 | UTR 3 | NP_001257639.1 | |||
NM_007243.2 | 1070 | Missense Mutation | CGC,TGC | R,C 240 | NP_009174.1 | |
XM_017010224.1 | 1070 | Missense Mutation | CGC,TGC | R,C 161 | XP_016865713.1 | |
XM_017010225.1 | 1070 | Missense Mutation | CGC,TGC | R,C 161 | XP_016865714.1 |
PPP1R18 - protein phosphatase 1 regulatory subunit 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134870.1 | 1070 | Intron | NP_001128342.1 | |||
NM_133471.3 | 1070 | Intron | NP_597728.1 |