Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTAACCTGGCTTGGTGGAAGGC[G/A]CTGGATTTTGGGTATCACTCCATAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612944 | ||||||||||||||||||||
Literature Links: |
RNASET2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNASET2 - ribonuclease T2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003730.4 | 957 | Missense Mutation | CGC,TGC | R,C 184 | NP_003721.2 | |
XM_017011397.1 | 957 | Intron | XP_016866886.1 | |||
XM_017011398.1 | 957 | Intron | XP_016866887.1 | |||
XM_017011399.1 | 957 | UTR 3 | XP_016866888.1 |