Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCCTGTGCCCCGGGGTCGGC[A/C]GTCTCCGTTACCTGGTTGGGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109170 MIM: 153440 MIM: 600978 MIM: 611550 MIM: 191160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287329 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100287329 - uncharacterized LOC100287329 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LST1 - leukocyte specific transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTA - lymphotoxin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTB - lymphotoxin beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002341.1 | 182 | Silent Mutation | ACG,ACT | T,T 58 | NP_002332.1 | |
NM_009588.1 | 182 | Intron | NP_033666.1 |
NCR3 - natural cytotoxicity triggering receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNF - tumor necrosis factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |