Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGGTTGATGCTGGTGAGTGTG[G/T]TGTTGATGGCGAGCACCTGTGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186975 | ||||||||||||||||||||
Literature Links: |
TCTE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCTE1 - t-complex-associated-testis-expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182539.3 | 1861 | Missense Mutation | AAC,ACC | N,T 418 | NP_872345.2 | |
XM_005248872.1 | 1861 | Missense Mutation | AAC,ACC | N,T 418 | XP_005248929.1 | |
XM_005248873.2 | 1861 | Intron | XP_005248930.1 | |||
XM_005248874.2 | 1861 | Missense Mutation | AAC,ACC | N,T 319 | XP_005248931.1 | |
XM_006715009.3 | 1861 | Missense Mutation | AAC,ACC | N,T 319 | XP_006715072.1 | |
XM_011514336.2 | 1861 | Missense Mutation | AAC,ACC | N,T 418 | XP_011512638.1 | |
XM_011514338.2 | 1861 | Missense Mutation | AAC,ACC | N,T 418 | XP_011512640.1 | |
XM_011514339.2 | 1861 | Missense Mutation | AAC,ACC | N,T 319 | XP_011512641.1 | |
XM_011514340.1 | 1861 | Intron | XP_011512642.1 |
TMEM151B - transmembrane protein 151B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |