Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAATAGGTCGTGGAGACTCTTCA[C/G]AGGCTGCTGTGTGAATCCCAGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614137 | ||||||||||||||||||||
Literature Links: |
FAM26E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM26E - family with sequence similarity 26 member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153711.3 | 285 | Missense Mutation | ACA,AGA | T,R 78 | NP_714922.1 |
TRAPPC3L - trafficking protein particle complex 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139444.2 | 285 | Intron | NP_001132916.1 |