Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGACCACGGGAGAAGACGAAT[A/G]TCGGGGAGGCCGTGGGGTCTGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605091 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGD2 - FYVE, RhoGEF and PH domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173558.3 | 332 | Missense Mutation | ATC,GTC | I,V 57 | NP_775829.2 | |
XM_011514372.2 | 332 | Missense Mutation | ATC,GTC | I,V 57 | XP_011512674.1 | |
XM_011514373.2 | 332 | Missense Mutation | ATC,GTC | I,V 57 | XP_011512675.1 | |
XM_017010429.1 | 332 | Missense Mutation | ATC,GTC | I,V 57 | XP_016865918.1 |