Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTACCTGCAACATTTCTCTTCTT[G/T]GTTTTCACAGCACGTTTTAGATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613973 | ||||||||||||||||||||
Literature Links: |
MB21D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MB21D1 - Mab-21 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138441.2 | 1347 | Missense Mutation | NP_612450.2 | |||
XM_017010232.1 | 1347 | Missense Mutation | XP_016865721.1 |