Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATGTGAGCTGCCAAGGAGGGT[C/G]AAGAGGAGGACAAGGGGCAGCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613928 MIM: 612802 | ||||||||||||||||||||
Literature Links: |
DPCR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPCR1 - diffuse panbronchiolitis critical region 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFTA2 - surfactant associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205854.2 | 112 | Missense Mutation | TTC,TTG | F,L 11 | NP_995326.1 |
VARS2 - valyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |