Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGTTTATCAAATCATACAGTG[A/G]TGGTAGTCTTATTTTATCCATATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35B3 - solute carrier family 35 member B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142541.2 | 1359 | Missense Mutation | CCA,TCA | P,S 380 | NP_001136013.1 | |
NM_015948.4 | 1359 | Missense Mutation | CCA,TCA | P,S 380 | NP_057032.2 | |
XM_005249156.3 | 1359 | Missense Mutation | CCA,TCA | P,S 382 | XP_005249213.1 | |
XM_005249157.2 | 1359 | Missense Mutation | CCA,TCA | P,S 380 | XP_005249214.1 | |
XM_005249158.1 | 1359 | Missense Mutation | CCA,TCA | P,S 348 | XP_005249215.1 | |
XM_006715102.3 | 1359 | Missense Mutation | CCA,TCA | P,S 380 | XP_006715165.1 | |
XM_006715103.3 | 1359 | Missense Mutation | CCA,TCA | P,S 351 | XP_006715166.1 | |
XM_011514648.1 | 1359 | Missense Mutation | CCA,TCA | P,S 348 | XP_011512950.1 | |
XM_011514650.2 | 1359 | Intron | XP_011512952.1 | |||
XM_017010910.1 | 1359 | Missense Mutation | CCA,TCA | P,S 380 | XP_016866399.1 | |
XM_017010911.1 | 1359 | Missense Mutation | CCA,TCA | P,S 380 | XP_016866400.1 | |
XM_017010912.1 | 1359 | Missense Mutation | CCA,TCA | P,S 349 | XP_016866401.1 | |
XM_017010913.1 | 1359 | Missense Mutation | CCA,TCA | P,S 349 | XP_016866402.1 | |
XM_017010914.1 | 1359 | Missense Mutation | CCA,TCA | P,S 348 | XP_016866403.1 | |
XM_017010915.1 | 1359 | Missense Mutation | CCA,TCA | P,S 317 | XP_016866404.1 |