Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCTGCAAGAGGAGCTGGCCCG[C/T]AGCTTCTTCTGGCAGGTGCTGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIM1 - Pim-1 proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243186.1 | 865 | Silent Mutation | CGC,CGT | R,R 236 | NP_001230115.1 | |
NM_002648.3 | 865 | Silent Mutation | CGC,CGT | R,R 145 | NP_002639.1 |