Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTTCCTCGTCGATGAAATCAGT[A/G]TCTATGACATTGGGCGGCAGCATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602937 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CITED2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CITED2 - Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168388.2 | 903 | Silent Mutation | GAC,GAT | D,D 219 | NP_001161860.1 | |
NM_001168389.2 | 903 | Silent Mutation | GAC,GAT | D,D 224 | NP_001161861.2 | |
NM_006079.4 | 903 | Silent Mutation | GAC,GAT | D,D 219 | NP_006070.2 |