Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGCAGGCAGCTCAGTTGGACA[A/C]GGAGGGTGGCAGGCCAGGACGCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604876 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCAN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCAN2 - regulator of calcineurin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251973.1 | 921 | Missense Mutation | NP_001238902.1 | |||
NM_001251974.1 | 921 | Missense Mutation | NP_001238903.1 | |||
NM_005822.3 | 921 | Missense Mutation | NP_005813.2 | |||
XM_011514226.1 | 921 | Missense Mutation | XP_011512528.1 |