Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606520 MIM: 602872 MIM: 604744 MIM: 610435 MIM: 606038 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf25 - chromosome 6 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025260.3 | 828 | Intron | NP_079536.2 | |||
NM_138272.2 | 828 | Intron | NP_612116.1 | |||
NM_138273.2 | 828 | Intron | NP_612117.1 | |||
NM_138274.2 | 828 | Intron | NP_612118.1 | |||
NM_138275.2 | 828 | Intron | NP_612119.1 | |||
NM_138277.2 | 828 | Intron | NP_612121.1 | |||
XM_011514920.2 | 828 | Intron | XP_011513222.1 | |||
XM_011514921.2 | 828 | Intron | XP_011513223.1 | |||
XM_017011330.1 | 828 | Intron | XP_016866819.1 | |||
XM_017011331.1 | 828 | Intron | XP_016866820.1 | |||
XM_017011332.1 | 828 | Intron | XP_016866821.1 | |||
XM_017011333.1 | 828 | Intron | XP_016866822.1 |
CLIC1 - chloride intracellular channel 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDAH2 - dimethylarginine dimethylaminohydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303007.1 | 828 | Silent Mutation | GCC,GCT | A,A 208 | NP_001289936.1 | |
NM_001303008.1 | 828 | Silent Mutation | GCC,GCT | A,A 208 | NP_001289937.1 | |
NM_013974.2 | 828 | Silent Mutation | GCC,GCT | A,A 208 | NP_039268.1 | |
XM_011514448.2 | 828 | Silent Mutation | GCC,GCT | A,A 208 | XP_011512750.1 |
LY6G6C - lymphocyte antigen 6 complex, locus G6C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G6D - lymphocyte antigen 6 complex, locus G6D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |