Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTCAATGTATTTGGAGATGCC[G/T]GTTCAAAAACGTCATCATCATCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604577 | ||||||||||||||||||||
Literature Links: |
BVES PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BVES - blood vessel epicardial substance | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199563.1 | 1135 | Missense Mutation | NP_001186492.1 | |||
NM_007073.4 | 1135 | Missense Mutation | NP_009004.2 | |||
NM_147147.3 | 1135 | Missense Mutation | NP_671488.1 | |||
XM_011535398.2 | 1135 | Missense Mutation | XP_011533700.1 |