Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTTTGGTCTGTATTTTCTGCAC[C/T]TCTTCAGGCCAAGAATAACAATCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 154250 | ||||||||||||||||||||
Literature Links: |
ME1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ME1 - malic enzyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002395.5 | 1951 | Silent Mutation | GAA,GAG | E,E 562 | NP_002386.1 | |
XM_011535836.2 | 1951 | Silent Mutation | GAA,GAG | E,E 487 | XP_011534138.1 |