Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGATTGAGGTAGAGGGGCTGGT[A/G]GGAGGCCTGGTGGGCCTGGCTGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600984 MIM: 600985 | ||||||||||||||||||||
Literature Links: |
ATF6B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATF6B - activating transcription factor 6 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136153.1 | 2116 | Missense Mutation | CAC,TAC | H,Y 692 | NP_001129625.1 | |
NM_004381.4 | 2116 | Missense Mutation | CAC,TAC | H,Y 695 | NP_004372.3 |
TNXB - tenascin XB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |