Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGTATCAGGATCCAAGGAACGT[C/T]TGGGGTTTCCTAGGTATGTCTGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM14B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM14B - transmembrane protein 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127711.2 | 356 | Intron | NP_001121183.1 | |||
NM_001286484.1 | 356 | Intron | NP_001273413.1 | |||
NM_001286488.1 | 356 | Silent Mutation | GTC,GTT | V,V 63 | NP_001273417.1 | |
NM_001286489.1 | 356 | Intron | NP_001273418.1 | |||
NM_030969.4 | 356 | Intron | NP_112231.3 |