Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTCGGCGCATTACGACGCCGT[A/G]GAGGCCGAGCTGAAGTCCAGCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614686 | ||||||||||||||||||||
Literature Links: |
FAM50B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM50B - family with sequence similarity 50 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012135.2 | 312 | Silent Mutation | GTA,GTG | V,V 56 | NP_036267.1 | |
XM_017010729.1 | 312 | Silent Mutation | GTA,GTG | V,V 56 | XP_016866218.1 |