Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCTTCTGTTTCCTCTGATTTTA[C/T]AAATTCCTTGATAAATTTCCAAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C6orf118 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf118 - chromosome 6 open reading frame 118 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144980.3 | 1604 | Missense Mutation | TAT,TGT | Y,C 462 | NP_659417.2 | |
XM_005266838.2 | 1604 | Missense Mutation | ATA,GTA | I,V 481 | XP_005266895.1 | |
XM_011535509.2 | 1604 | Missense Mutation | ATA,GTA | I,V 516 | XP_011533811.1 | |
XM_011535510.2 | 1604 | Missense Mutation | ATA,GTA | I,V 496 | XP_011533812.1 | |
XM_011535511.2 | 1604 | Missense Mutation | TAT,TGT | Y,C 497 | XP_011533813.1 | |
XM_011535512.2 | 1604 | Missense Mutation | ATA,GTA | I,V 377 | XP_011533814.1 | |
XM_011535513.2 | 1604 | Missense Mutation | ATA,GTA | I,V 377 | XP_011533815.1 | |
XM_017010323.1 | 1604 | Intron | XP_016865812.1 | |||
XM_017010324.1 | 1604 | Missense Mutation | ATA,GTA | I,V 377 | XP_016865813.1 |