Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCGGTAGCGATGGACACTCTGG[A/G]TCGAGTAGTGTAAGTGCGCTGGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RPF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPF2 - ribosome production factor 2 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289111.1 | 104 | UTR 5 | NP_001276040.1 | |||
NM_032194.2 | 104 | Missense Mutation | GAT,GGT | D,G 5 | NP_115570.1 | |
XM_017011354.1 | 104 | Intron | XP_016866843.1 |