Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCCATTCTCTGCATTTTGGTTG[C/T]GTGGATCTTTAAAAATGCCGACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612025 | ||||||||||||||||||||
Literature Links: |
IYD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IYD - iodotyrosine deiodinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164694.1 | 190 | Missense Mutation | GCG,GTG | A,V 17 | NP_001158166.1 | |
NM_001164695.1 | 190 | Missense Mutation | GCG,GTG | A,V 17 | NP_001158167.1 | |
NM_001318495.1 | 190 | UTR 5 | NP_001305424.1 | |||
NM_203395.2 | 190 | Missense Mutation | GCG,GTG | A,V 17 | NP_981932.1 | |
XM_006715478.3 | 190 | Missense Mutation | GCG,GTG | A,V 17 | XP_006715541.1 | |
XM_006715479.3 | 190 | Intron | XP_006715542.1 |