Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTCCCCTTGCAGAATGGCCA[A/G]AGTTTTCTGGTCTTGGATGAGCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140581 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSF2 - heat shock transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135564.1 | 289 | Silent Mutation | CAA,CAG | Q,Q 34 | NP_001129036.1 | |
NM_001243094.1 | 289 | Silent Mutation | CAA,CAG | Q,Q 34 | NP_001230023.1 | |
NM_004506.3 | 289 | Silent Mutation | CAA,CAG | Q,Q 34 | NP_004497.1 |