Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCCAGTGCACCAGCCTCAGCC[C/T]ACGGAGAAGAGTGACTGAGGCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604797 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC2 - apolipoprotein B mRNA editing enzyme catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPO2 - translocator protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010873.2 | 606 | Silent Mutation | CCC,CCT | P,P 165 | NP_001010873.1 | |
NM_001159726.1 | 606 | Silent Mutation | CCC,CCT | P,P 165 | NP_001153198.1 | |
XM_011514396.2 | 606 | Silent Mutation | CCC,CCT | P,P 165 | XP_011512698.1 | |
XM_011514397.2 | 606 | Silent Mutation | CCC,CCT | P,P 165 | XP_011512699.1 |
UNC5CL - unc-5 family C-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |