Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAGGTGGTCGCTGGAGAGGCA[C/T]CTCTAGGCCCCCGGAGGCCGTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX43 - DEAD-box helicase 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018665.2 | 505 | Missense Mutation | ACC,ATC | T,I 54 | NP_061135.2 | |
XM_011535926.1 | 505 | Missense Mutation | ACC,ATC | T,I 54 | XP_011534228.1 | |
XM_011535927.2 | 505 | Intron | XP_011534229.1 | |||
XM_011535928.2 | 505 | UTR 5 | XP_011534230.1 | |||
XM_017011003.1 | 505 | UTR 5 | XP_016866492.1 |
OOEP-AS1 - OOEP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |