Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGCAGCGGCCCTGTCTCCTCTG[A/G]GGGCCTTATGCAGCTGCCTACCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607489 MIM: 600589 | ||||||||||||||||||||
Literature Links: |
CUL9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL9 - cullin 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRF - serum response factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292001.1 | 1363 | Missense Mutation | GAG,GGG | E,G 130 | NP_001278930.1 | |
NM_003131.3 | 1363 | Missense Mutation | GAG,GGG | E,G 334 | NP_003122.1 |