Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCCCCTCTTACCATCTCCATT[A/C]TCATCCACCAGGAGGAAGATCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600364 MIM: 602275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCA1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCA1A - guanylate cyclase activator 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GUCA1B - guanylate cyclase activator 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002098.5 | 601 | Missense Mutation | GAG,GAT | E,D 155 | NP_002089.4 |